Learn More
Eppendorf™ Frame For Worktop
Product Code. 10324271 Shop All Eppendorf Products
Change view
Click to view available options
For Use With (Application):
All Thermal Cycler with 0.2mL block format
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Application) Unit Size
10324271 All Thermal Cycler with 0.2mL block format Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10324271 Supplier Eppendorf™ Supplier No. 0030124243
Estimated Shipment: 20-10-2026
A contamination shield on the front edge of the lid prevents accidental contact with the inner surface of the lid during opening, thus eliminating a serious contamination risk
A contamination shield on the front edge of the lid prevents accidental contact with the inner surface of the lid during opening, thus eliminating a serious contamination risk. The special lid hinge allows the lid to click into a set position of approximately 90° after the test tube has been opened.
  • Can be used with all thermal cyclers with 0.2mL block format
  • Contamination shield on lid
  • Special hinge for fixed lid position
  • High transparency
  • Certified PCR* clean
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) All Thermal Cycler with 0.2mL block format