missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FOX2/RBM9 Mouse anti-Human, Unconjugated, Clone: 2D7G5, Proteintech
Mouse Monoclonal Antibody
1710.33 NOK - 4836.10 NOK
Specifications
| Antigen | FOX2/RBM9 |
|---|---|
| Clone | 2D7G5 |
| Concentration | 820 μg/mL |
| Applications | Western Blot, Immunohistochemistry, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16849315
|
Proteintech
66976-1-IG-20UL |
20 μL |
1710.33 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16839315
|
Proteintech
66976-1-IG-150UL |
150 μL |
4836.10 NOK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The FOX2/RBM9 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human FOX2/RBM9. This antibody recognizes human, mouse antigen. The FOX2/RBM9 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| FOX2/RBM9 | |
| 820 μg/mL | |
| Monoclonal | |
| Mouse | |
| 23543 | |
| Primary |
| 2D7G5 | |
| Western Blot, Immunohistochemistry, ELISA | |
| Unconjugated | |
| Human, Mouse | |
| IgG1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title