Learn More
FOX2/RBM9 Mouse anti-Human, Unconjugated, Clone: 2D7G5, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66976-1-IG-150UL
Description
The FOX2/RBM9 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human FOX2/RBM9. This antibody recognizes human, mouse antigen. The FOX2/RBM9 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| FOX2/RBM9 | |
| Monoclonal | |
| 820 μg/mL | |
| Mouse | |
| Primary | |
| Human, Mouse |
| Western Blot, Immunohistochemistry, ELISA | |
| 2D7G5 | |
| Unconjugated | |
| 150 μL | |
| 23543 | |
| IgG1 |