Learn More
FOX2/RBM9 Mouse anti-Human, Unconjugated, Clone: 2D7G5, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 66976-1-IG-20UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16849315
  • 1415.13 NOK / 20µL
Estimated Shipment: 10-10-2023
Description
The FOX2/RBM9 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human FOX2/RBM9. This antibody recognizes human, mouse antigen. The FOX2/RBM9 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
FOX2/RBM9
Monoclonal
820 μg/mL
Mouse
Primary
Human, Mouse
Western Blot, Immunohistochemistry, ELISA
2D7G5
Unconjugated
20 μL
23543
IgG1