Description
- Used for microbiology applications
- During storage of media, it is preferable to cover the stoppers with aluminum foil to avoid dust contamination on removal
- Shrinkage will occur after continued use following autoclaving, but this is dependent purely on application
- To eliminate the possibility of obstructing the flow of air into the media container, care should be taken not to crease the stoppers on fitting
Specifications
Specifications
| Diameter (Metric) | 25mm |
| For Use With (Application) | Microbiology |
| Length (Metric) | 50 mm |
| Quantity | 100/Pk. |
| Size | 25 x 50mm |
| Color | White |
| Material | Polyurethane |
| Product Type | Stopper |