Learn More
Fisherbrand™ Foam Stand for Microtiter Tubes
Product Code. 11792583 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Color:
Black
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
11792583 Black Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11792583
  • ONLINE EXCLUSIVE PRICE
    459.00 NOK
    450.45 NOK / Pack of 1
    Save 8.55 NOK
    1% Off
Estimated Shipment: 18-12-2026
Add to basket
Add to basket
Securely holds microtiter tubes during the mixing process. Foam Stands for Microtiter Tubes are for use with Vortex Mixers.
  • Soft foam to prevent breakage
  • Plate not included
Specifications
Product Type Foam Stand
Color Black
For Use With (Equipment) CLASSIC, ZX3, WIZARD, ZX4, TX4 Vortex Mixers