missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FN3K Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14293-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
FN3K catalyzes phosphorylation of fructosamines formed by glycation, the nonenzymatic reaction of glucose with primary amines followed by Amadori rearrangement. Phosphorylation of fructosamines may initiate metabolism of the modified amine and result in deglycation of glycated proteins.Specifications
FN3K | |
Polyclonal | |
Unconjugated | |
FN3K | |
FN3K, fructosamine 3 kinase | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
63828, 64122 | |
-20°C | |
Liquid |
Western Blot, Immunofluorescence, Immunocytochemistry, Immunoprecipitation | |
0.19 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9ER35, Q9H479 | |
Fn3k | |
FN3K Fusion Protein Ag5617 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |