missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FN3K Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1611.84 NOK - 4593.76 NOK
Specifications
Antigen | FN3K |
---|---|
Concentration | 0.19 mg/mL |
Applications | Western Blot, Immunofluorescence, Immunocytochemistry, Immunoprecipitation |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16892864
|
Proteintech
14293-1-AP-20UL |
20 μL |
1611.84 NOK
20µL |
Estimated Shipment: 14-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16882864
|
Proteintech
14293-1-AP-150UL |
150 μL |
4593.76 NOK
150µL |
Estimated Shipment: 14-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
FN3K catalyzes phosphorylation of fructosamines formed by glycation, the nonenzymatic reaction of glucose with primary amines followed by Amadori rearrangement. Phosphorylation of fructosamines may initiate metabolism of the modified amine and result in deglycation of glycated proteins.Specifications
FN3K | |
Western Blot, Immunofluorescence, Immunocytochemistry, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Mouse, Human | |
Q9ER35, Q9H479 | |
63828, 64122 | |
FN3K Fusion Protein Ag5617 | |
Primary | |
-20°C | |
FN3K |
0.19 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
FN3K, fructosamine 3 kinase | |
Fn3k | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |