Learn More
Applied Biosystems™ Fmoc-L-Phenylalanine, 10/Pk.
Product Code. 15802644 Shop All Applied Biosystems Products
Change view
Click to view available options
433A Peptide Synthesizer
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
15802644 433A Peptide Synthesizer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15802644 Supplier Applied Biosystems™ Supplier No. 410645
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
These are Amino Acid Cartridges for the ABI™ 431-433 Peptide Synthesizer.
  • For Research Use Only
  • Not for use in diagnostics procedures
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Each package contains 10 cartridges.
Format Cartridge
For Use With (Equipment) 433 A Peptide Synthesizer
For Use With (Application) Protein Sequencing & Peptide Synthesis
Product Type Peptide Synthesis Reagents
Quantity 10 Ea.