Learn More
WTW™ Flow-through Vessel for WTW™ pH Electrode
Specifications
Specifications
| Capacity (Metric) | 97 mL |
| Color | Transparent |
| Description | Flow-through vessel |
| For Use With (Equipment) | pH Electrodes, LTA 01 and LR 325/01 Conductivity Cells, TFK 150 and TFK 530 Temperature Probes |
| For Use With (Application) | For ultrapure water measuring |
| Material | PVC |