missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FLI1 Antibody (FLI1/1312), Novus Biologicals™
Mouse Monoclonal Antibody
2685.00 NOK - 5135.00 NOK
Specifications
| Antigen | FLI1 |
|---|---|
| Clone | FLI1/1312 |
| Concentration | 0.2 mg/mL |
| Dilution | Western Blot 1 - 2 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml |
| Applications | Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18149539
|
Novus Biologicals
NBP2-53335-20UG |
20 μg |
2685.00 NOK
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18179279
|
Novus Biologicals
NBP2-53335-100UG |
100 μg |
5135.00 NOK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FLI1 Monoclonal specifically detects FLI1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
| FLI1 | |
| 0.2 mg/mL | |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Cancer, Tumor Suppressors | |
| 10mM PBS with 0.05% BSA with 0.05% Sodium Azide | |
| 2313 | |
| Recombinant fragment (aa 107-261) of human FLI1 protein (exact sequence is proprietary) | |
| Primary | |
| Store at 4C. |
| FLI1/1312 | |
| Western Blot 1 - 2 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| EWSR2, Friend leukemia integration 1 transcription factor, Friend leukemia virus integration 1, Proto-oncogene Fli-1, SIC-1, Transcription factor ERGB | |
| FLI1 | |
| IgG | |
| Protein A purified | |
| 51 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title