missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FLI1 Antibody (FLI1/1312), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-53335-100UG
This item is not returnable.
View return policy
Description
FLI1 Monoclonal specifically detects FLI1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications
| FLI1 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Western Blot 1 - 2 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml | |
| EWSR2, Friend leukemia integration 1 transcription factor, Friend leukemia virus integration 1, Proto-oncogene Fli-1, SIC-1, Transcription factor ERGB | |
| Mouse | |
| 51 kDa | |
| 100 μg | |
| Cancer, Tumor Suppressors | |
| 2313 | |
| Store at 4C. | |
| IgG |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| FLI1/1312 | |
| Unconjugated | |
| 10mM PBS with 0.05% BSA with 0.05% Sodium Azide | |
| FLI1 | |
| Recombinant fragment (aa 107-261) of human FLI1 protein (exact sequence is proprietary) | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction