Learn More
Proteintech™ FlexAble CoraLite™ Plus 650 Antibody Labeling Kit for Rabbit IgG
Product Code. 17701519 Shop All Proteintech Products
Click to view available options
Quantity:
10 x 0.5 μg Reactions
200 x 0.5 μg Reactions
50 x 0.5 μg Reactions
Unit Size:
10 reactions
200 reactions
50 reactions
Product Code. 17701519
Brand: Proteintech™ KFA00310
This Antibody Labeling Kit conjugates CoraLite™ 650 into Rabbit IgG antibodies.
FlexAble CoraLite™ Plus 650 Antibody Labeling Kit for Rabbit IgG is a novel antibody labeling kit that uses an affinity linker to conjugate CoraLite™ Plus 650 in any buffer condition to rabbit IgG primary antibodies from any supplier.
  • One labeling reaction requires only 0.5 μg of antibody, regardless of the antibody concentration
  • Label your rabbit IgG antibody in <10 min without any additional equipment
Specifications
Kit Contents 10 μL CoraLite™ Plus 650 FlexLinker for Rabbit IgG, 20 μL FlexQuencher for Rabbit IgG, 100 μL FlexBuffer, Manual, Protocol Magnet
Quantity 10 x 0.5 μg Reactions
Content And Storage 1 Year at -20°C or 6 Months at +4°C, Avoid Light
Excitation/Emission 654 nm/674 nm
Labeling Target IgG
Species Rabbit