missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FKBP14 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15884-1-AP-150UL
This item is not returnable.
View return policy
Description
PPIases accelerate the folding of proteins during protein synthesis.Specifications
| FKBP14 | |
| Polyclonal | |
| Unconjugated | |
| FKBP14 | |
| 22 kDa FK506 binding protein, 22 kDa FKBP, FK506 binding protein 14, FKBP 14, FKBP 22, FKBP14, FKBP22, PPIase FKBP14, Rotamase | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 231997, 362366, 55033 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.41 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P59024, Q9NWM8 | |
| Fkbp14 | |
| FKBP14 Fusion Protein Ag8668 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction