missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FKBP14 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1710.33 NOK - 4836.10 NOK
Specifications
| Antigen | FKBP14 |
|---|---|
| Concentration | 0.41 mg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16855274
|
Proteintech
15884-1-AP-20UL |
20 μL |
1710.33 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16845274
|
Proteintech
15884-1-AP-150UL |
150 μL |
4836.10 NOK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PPIases accelerate the folding of proteins during protein synthesis.Specifications
| FKBP14 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Rat, Human, Mouse | |
| P59024, Q9NWM8 | |
| 231997, 362366, 55033 | |
| FKBP14 Fusion Protein Ag8668 | |
| Primary | |
| -20°C | |
| FKBP14 |
| 0.41 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| 22 kDa FK506 binding protein, 22 kDa FKBP, FK506 binding protein 14, FKBP 14, FKBP 22, FKBP14, FKBP22, PPIase FKBP14, Rotamase | |
| Fkbp14 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title