Learn More
Eppendorf™ Fixed Angle QuickLock™ Rotor with Aerosol-tight Lid
Shop All Eppendorf Products
Click to view available options
Capacity:
24 Spin Columns
30 x 1.5/2 mL
48 x 1.5 mL
6 x 15/50 mL
Unit Size:
Each
Description
- Aerosol-tight Eppendorf QuickLock rotors close with only 1/4 of a turn
- Saves time during every day use
- Takes repetitive stress off your wrist
Specifications
Specifications
Description | Model F-45-48-11 |
Type | Fixed Angle Rotor |
Capacity | 30 x 1.5/2 mL |
Max. RCF | 20,800 x g |
Max. Speed | 14,000 rpm |
For Use With (Equipment) | 5430/5430R Centrifuges |
Angle | Fixed |
Includes | Aerosol rotor lid |
No. of Places | 30 |
Promotions
-
Multiple Manufacturers
Equipment End of Year Sale
Don't miss our end-of-year sale: save up to 50% on premium lab equipment, instruments and services.Promo Code: 24121Expires: 2025-12-20
Product Content Correction
Product Title
Eppendorf™ Fixed Angle QuickLock™ Rotor with Aerosol-tight Lid >