Learn More
Fisherbrand™ Weighing Funnels
Shop All Fisherbrand Products
Click to view available options
Diameter (Metric)Top:
25 mm
Unit Size:
Each
Pack of 5
Specifications
Specifications
| Diameter (Metric)Top | 25 mm |
| Diameter (Metric) Stem | 10.25mm (OD); 8.12mm (ID) |
| Material | Borosilicate Glass |
| Height (Metric) | 55 mm |
| For Use With (Application) | Used for microtransfer of solids and liquids |
| Max. Temperature (English) | 849°F |
| Max. Temperature (Metric) | 454°C |
| Product Type | Weighing Funnel |
Product Content Correction
Product Title
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering