Description
- Multiple prongs for to support multiple tools
- Dual handles easily secures holder in the ultrasonic cleaner
- Stainless steel
- Light and easy to maneuver
Specifications
Specifications
| Type | Tool Holder |
| For Use With (Equipment) | Elmasonic EP20H E-Plus Ultrasonic Cleaner (107-1653), or the FB11203 Advanced Ultrasonic Cleaner |
| Height (English) | 3.6 in. |
| Length (English) | Bottom L: 4.5 in.; Top L: 8.2 in. |
| Material | Stainless Steel |
| Width (English) | Bottom W: 4.5 in.; Top W: 3 in. |