Learn More
- Home
- Products
- Stoppers and Closures
- Stoppers
- Fisherbrand™ Turnover Septum Stoppers
Fisherbrand™ Turnover Septum Stoppers
Product Code. 11512493 Shop All Fisherbrand Products| Product Code. | Diameter (Metric) Outer | Diameter (Metric) Inner | Color | unitSize |
|---|---|---|---|---|
| 11512493 | 13 mm | 8 mm | Red | Pack of 10 |
| 11756477 | 9 mm | 6.5 mm | Red | Pack of 10 |
| 11502493 | 9 mm | 6.5 mm | Red | Pack of 10 |
| 11766477 | 13 mm | 8 mm | Red | Pack of 10 |
| 11776477 | 17 mm | 9.5 mm | Red | Pack of 10 |
| 11532493 | 17 mm | 9.5 mm | Red | Pack of 10 |
| 11786477 | 21 mm | 11 mm | Red | Pack of 10 |
| 11542493 | 21 mm | 11 mm | Red | Pack of 10 |
| 11706487 | 33 mm | 16 mm | Red | Pack of 10 |
| 11572493 | 33 mm | 16 mm | Red | Pack of 10 |
| 11716477 | 29 mm | 14 mm | Red | Pack of 10 |
| 11562493 | 29 mm | 14 mm | Red | Pack of 10 |
| 10709984 | 25 mm | 12.5 mm | Red | Pack of 10 |
| 11552493 | 25 mm | 12.5 mm | Red | Pack of 10 |
| 11746477 | 41 mm | 19 mm | Red | Pack of 10 |
| 11592493 | 41 mm | 19 mm | Red | Pack of 10 |
| 11716487 | 45 mm | 20 mm | Red | Pack of 10 |
| 11502503 | 45 mm | 20 mm | Red | Pack of 10 |
| 11532503 | 57 mm | 25 mm | Red | Pack of 10 |
| 10171552 | 57 mm | 25 mm | Red | Pack of 10 |
| 11522503 | 53 mm | 24 mm | Red | Pack of 10 |
| 11588122 | 17 mm | 9.5 mm | Cream | Pack of 10 |
| 11578122 | 13 mm | 8 mm | Cream | Pack of 10 |
| 11598122 | 21 mm | 11 mm | Cream | Pack of 10 |
| 11518132 | 29 mm | 14 mm | Cream | Pack of 10 |
| 10777864 | 29 mm | 14 mm | Cream | Pack of 10 |
| 11508132 | 25 mm | 12.5 mm | Cream | Pack of 10 |
| 11528132 | 33 mm | 16 mm | Cream | Pack of 10 |
| 11538132 | 37 mm | 17.5 mm | Cream | Pack of 10 |
| 11548132 | 41 mm | 19 mm | Cream | Pack of 10 |
| 11558132 | 45 mm | 20 mm | Cream | Pack of 10 |
| 11568132 | 49 mm | 22 mm | Cream | Pack of 10 |
| 11578132 | 53 mm | 24 mm | Cream | Pack of 10 |
| 11588132 | 57 mm | 25 mm | Cream | Pack of 10 |
| 11764306 | 37mm | 17.5 mm | Red | Pack of 10 |
| 11582493 | 37mm | 17.5 mm | Red | Pack of 10 |
| 11786457 | 49 mm | 6.5 mm | Red | Pack of 10 |
| 11512503 | 49 mm | 6.5 mm | Red | Pack of 10 |
| 11568122 | 9 mm | 6.5 mm | Cream | Pack of 10 |
Product Code. 11512493
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Specifications
| Diameter (Metric) Inner | 8 mm |
| For Use With (Application) | For narrow aperture containers |
| For Use With (Equipment) | For 0.92 to 0.97cm I.D. containers |
| Total Positions | 1 |
| Color | Red |
| Diameter (Metric) Outer | 13 mm |
| Material | Rubber |
| Product Type | Turnover Septum Stopper |