Learn More
Fisherbrand™ Sterile Microcentrifuge Tubes with Screw Caps
Product Code. 12350353 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Capacity (Metric):
1.5 mL
2 mL
Max. RCF:
18,000 x g
25,000 x g
30,000 x g
Unit Size:
Pack of 500
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Max. RCF Unit Size
12350353 1.5 mL 30,000 x g Pack of 500
12831731 2 mL 18,000 x g Pack of 500
12390433 2 mL 25,000 x g Pack of 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 12350353
Estimated Shipment: 10-12-2026
Add to basket
Add to basket
Note: Image is representative only. It shows colored caps, but all of the tubes in this offering come with Natural caps; there are no color cap options.
Choose skirted or conical-bottom style
  • With attached caps
  • Natural-colored screw caps with O-rings
  • RNase-free and DNase-free
TRUSTED_SUSTAINABILITY
Specifications
Closure Material Polypropylene
Autoclavable Autoclavable
Capacity (Metric) 1.5 mL
Color Natural
Material Polypropylene
Includes O-ring
Sterility Sterile
Closure Type Screw Cap
Bottom Shape Conical
Max. RCF 30,000 x g