Learn More
Fisherbrand™ Shipping Labels
Product Code. p-192659 Shop All Fisherbrand Products
Change view
Click to view available options
Product Type:
Dry Ice Mini Super Label
Size (English):
6 x 6 in.
Size (Metric):
15.24 x 15.24 cm
Unit Size:
Pack of 12
Product Code. Product Type Size (English) Size (Metric) unitSize
16478115 Dry Ice Mini Super Label 6 x 6 in. 15.24 x 15.24 cm Pack of 12
1 options
Product Code. 16478115
Only null left
Add to basket
Add to basket
For shipping infectious substances and diagnostic specimens
Specifications
Format Label
Product Type Dry Ice Mini Super Label
For Use With (Application) Shipping
Quantity 12/Pk.
Size (English) 6 x 6 in.
Size (Metric) 15.24 x 15.24 cm