Learn More
Fisherbrand™ Shipping Labels
Product Code. 16478115 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Product Type:
Dry Ice Mini Super Label
Size (English):
6 x 6 in.
Unit Size:
Pack of 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Size (English) Unit Size
16478115 Dry Ice Mini Super Label 6 x 6 in. Pack of 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16478115
Stock Up and Save Up to 60%
Use promo code 28009
View all promotions
Estimated Shipment: 29-12-2026
Add to basket
Add to basket
For shipping infectious substances and diagnostic specimens
Specifications
Format Label
Product Type Dry Ice Mini Super Label
For Use With (Application) Shipping
Quantity 12/Pk.
Size (English) 6 x 6 in.
Size (Metric) 15.24 x 15.24 cm