Learn More
Fisherbrand™ Round Cuvets
Product Code. p-81936 Shop All Fisherbrand Products
Change view
Click to view available options
Diameter (Metric):
19 mm
Height (Metric):
105 mm
Unit Size:
Pack of 36
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Height (Metric) unitSize
16333416 19 mm 105 mm Pack of 36
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16333416
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
For use with commercial colorimeters and spectrophotometers.
  • Reference mark near top assists repositioning
  • Manufactured from borosilicate glass
  • Available in case of 36
Specifications
Type Cuvette
Material Borosilicate Glass
Height (Metric) 105 mm
For Use With (Equipment) Coleman™ Jr. II
Shape Round
Diameter (Metric) 19 mm