Description
- Rotary Mixer Disc complete with clips for tubes, supplied unassembled
- Suitable for 'End to End Mixing' tubes,
- Clips for tube diameters between 8 - 11mm are suitable for 0.5mL, 1.5mL and 2.0mL Microcentrifuge tubes, 3mL Vacutainers and 5mL capped tubes (8 -11mm)
- Clips for tube diameters between 15 - 17mm are suitable for 15mL centrifuge tubes
- Easy to fit onto the RM-1 locating on three pins then secured in place with thumb wheel
Specifications
Specifications
| Product Type | Rotary Mixer Disc |
| For Use With (Equipment) | 8-11 mm dia. Tubes |