Learn More
Fisherbrand™ Reusable Polyethylene Dippers
Product Code. 16572440 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Length (Metric) Handle:
1.8 m
Length (English) Handle:
6 ft.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Handle Length (English) Handle Unit Size
16572440 1.8 m 6 ft. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16572440
Top Sellers. Mega Savings.
Use promo code 27997
View all promotions
Estimated Shipment: 04-01-2027
Add to basket
Add to basket
500mL beakers with long handles for removing samples from large tanks.
  • Lightweight, durable polyethylene
  • Graduated beaker features dual pouring spouts for easy left- or right-handed operation
  • Handle has an “easy-grip” end for secure handling under adverse conditions
Order Info
Additional dipper sizes available. To order, contact your Fisher Scientific Customer Service Representative.
Specifications
Product Type Dipper
Material HDPE
Capacity (Metric) 500 mL
For Use With (Application) For removing samples from large tanks
Dimensions (L x W x H) 78.5 x 5 x 4.25 in. (199.4 x 12.7 x 10.79 cm)
Height (English) 4.25 in.
Height (Metric) 10.79 cm
Length (English) 78.5 in.
Length (English) Handle 6 ft.
Length (Metric) 199.4 cm
Length (Metric) Handle 1.8 m
Width (English) 5 in.
Width (Metric) 12.7 cm