Learn More
Fisherbrand™ Powder Funnels
GREENER_CHOICE
Product Code. p-129565 Shop All Fisherbrand Products
Change view
Click to view available options
Diameter (Metric)Top:
6.5cm
7.5cm
10.5cm
15cm
Diameter (Metric) Stem:
16mm
21mm
27mm
Capacity (Metric):
50 mL
100 mL
225 mL
750 mL
Height (Metric):
6.6 cm
8.4 cm
10.3 cm
13.7 cm
Unit Size:
Pack of 12
Pack of 24
Pack of 36
Pack of 4
Pack of 6
Product Code. Diameter (Metric)Top Diameter (Metric) Stem Capacity (Metric) Height (Metric) unitSize
12652435 15cm 27mm 750 mL 13.7 cm Pack of 4
11907914 15cm 27mm 750 mL 13.7 cm Pack of 24
11997904 10.5cm 21mm 225 mL 10.3 cm Pack of 24
12642435 10.5cm 21mm 225 mL 10.3 cm Pack of 6
11907924 7.5cm 16mm 100 mL 8.4 cm Pack of 36
12632435 7.5cm 16mm 100 mL 8.4 cm Pack of 12
12622435 6.5cm 16mm 50 mL 6.6 cm Pack of 12
11997914 6.5cm 16mm 50 mL 6.6 cm Pack of 36
8 options
Product Code. 12652435
Only null left
Add to basket
Edge
Add to basket
Parallel stems minimize bridging of powder.
Parallel stems minimize bridging of powder
  • Durable polypropylene
  • External ribbing prevents air locks
  • Autoclavable
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric)Top 15cm
Capacity (Metric) 750 mL
Diameter (Metric) Stem 27mm
Material Polypropylene
Height (Metric) 13.7 cm
Autoclavable Autoclavable
Length (Metric) Stem 30mm