Learn More
Fisherbrand™ Powder Funnels
GREENER_CHOICE
Product Code. 12642435 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Diameter (Metric)Top:
6.5cm
7.5cm
10.5cm
15cm
Diameter (Metric) Stem:
16mm
21mm
27mm
Capacity (Metric):
50 mL
100 mL
225 mL
750 mL
Height (Metric):
6.6 cm
8.4 cm
10.3 cm
13.7 cm
Unit Size:
Pack of 12
Pack of 24
Pack of 36
Pack of 4
Pack of 6
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric)Top Diameter (Metric) Stem Capacity (Metric) Height (Metric) Unit Size
12642435 10.5cm 21mm 225 mL 10.3 cm Pack of 6
12652435 15cm 27mm 750 mL 13.7 cm Pack of 4
11907914 15cm 27mm 750 mL 13.7 cm Pack of 24
11997904 10.5cm 21mm 225 mL 10.3 cm Pack of 24
12632435 7.5cm 16mm 100 mL 8.4 cm Pack of 12
11907924 7.5cm 16mm 100 mL 8.4 cm Pack of 36
12622435 6.5cm 16mm 50 mL 6.6 cm Pack of 12
11997914 6.5cm 16mm 50 mL 6.6 cm Pack of 36
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 12642435
In Stock
Add to basket
Edge
Add to basket
Parallel stems minimize bridging of powder.
Parallel stems minimize bridging of powder
  • Durable polypropylene
  • External ribbing prevents air locks
  • Autoclavable
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric)Top 10.5cm
Capacity (Metric) 225 mL
Diameter (Metric) Stem 21mm
Material Polypropylene
Height (Metric) 10.3 cm
Autoclavable Autoclavable
Length (Metric) Stem 33mm