Learn More
Fisherbrand™ Porcelain Crucible Covers
Product Code. 13449709 Shop All Fisherbrand Products
Change view
Click to view available options
Diameter (Metric) Outer:
30mm
40mm
44mm
53mm
60mm
75mm
87mm
99mm
115mm
Diameter (Metric) Inner:
26mm
34mm
38mm
45mm
54mm
66mm
77mm
90mm
105mm
Height (Metric):
5mm
5.5mm
6mm
7mm
8mm
9mm
Thickness (Metric):
1.75mm
1.9mm
2.3mm
2.5mm
2.75mm
3mm
Unit Size:
Pack of 6
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Outer Diameter (Metric) Inner Height (Metric) Thickness (Metric) unitSize
13449709 53mm 45mm 7mm 2.3mm Pack of 6
13419709 30mm 26mm 5mm 1.75mm Pack of 6
13439709 40mm 34mm 5.5mm 1.75mm Pack of 6
13429709 44mm 38mm 6mm 1.9mm Pack of 6
13469709 75mm 66mm 7mm 2.5mm Pack of 6
13459709 60mm 54mm 7mm 2.5mm Pack of 6
13479709 87mm 77mm 8mm 2.75mm Pack of 6
13499709 115mm 105mm 9mm 3mm Pack of 6
13489709 99mm 90mm 8mm 2.75mm Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 13449709
For use with Fisherbrand High- and Wide-Form Porcelain Crucibles.
For use with Fisherbrand High- and Wide-Form Porcelain Crucibles
  • Glazed inside and out except for the outside bottom surface
  • With perforated handle
  • Strongly recommended that heating and cooling rates not exceed 200°C per hour and maximum use temperature of 1100°C
Specifications
Description With Perforated handle
Diameter (Metric) Inner 45mm
Material Porcelain
Finish Glazed
For Use With (Application) Use with Fisherbrand high and wide form porcelain crucible
Max. Temperature (Metric) 1150°C
Thickness (Metric) 2.3mm
Weight (Metric) 24g
Diameter (Metric) Outer 53mm
Height (Metric) 7mm
Product Type Crucible Cover