Description
Fisherbrand™ Permanent Marking Pens write on glass, film, wood, plastic, metal, rubber, and porcelain. Perfect for marking sample bags, microscope or photographic slides, laboratory glassware and more. Write on all surfaces.
- Produce precise, ultrathin lines 0.3mm wide
- Waterproof and fadeproof, permanent ink dries almost instantly
- An airtight ring seal cap prevents dry-out and ensures instant writing even after lengthy storage
- Unique stainless-steel long leg allows precise writing in a confined space
Order Info
Assorted includes six opaque black, two vivid red, two blue, and two green pens
Specifications
Specifications
| Tip Style | 0.3 mm Ultra Fine |
| Product Type | Permanent Pen |
| Color | Blue |
| Material | Stainless Steel |
| For Use With (Application) | Perfect marking pens for lab samples, microscope slides, photographic slides, glass and plastic beakers, and plastic sample bags |
| Quantity | 6 Pk. |
| Width (Metric) Line | 0.3 mm |