Learn More
Fisherbrand™ Multiple-Roll Tape Dispenser
GREENER_CHOICE
Product Code. p-190836 Shop All Fisherbrand Products
Change view
Click to view available options
Holds:
Tapes with 1 in. Core
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Holds unitSize
10092594 Tapes with 1 in. Core Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10092594
Only null left
Add to basket
Edge
Add to basket
A multiple tape dispenser provides a well-organized means of storing a rainbow assortment of color coding tape.
  • For dispensing multiple rolls with a 1 in. dia. (76.2mm) core (14 yd. [13m] of tape)
  • Accommodate 0.50, 0.75, or 3 in. wide (13, 19, or 25mm) tapes for a total dispensing width of 9.50 in. (24cm)
  • Dispensers stay put with a rubbery felt-type material to keep them from sliding
Order Info
Order tapes separately.
Specifications
Product Type Multiple-Roll Tape Dispenser
Holds Tapes with 1 in. Core
Material Metal
For Use With (Equipment) For dispensing 9.5 W, rolls with 14 yd. tape