Learn More
Fisherbrand™ Infecon™ 2000 and 2050 Infectious/Diagnostic Shippers
Product Code. 15502066 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Type:
Infecon 2000 Shipper
Size:
4 x 4 x 8 in.
Unit Size:
Pack of 6
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Size Unit Size
15502066 Infecon 2000 Shipper 4 x 4 x 8 in. Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15502066
  • ONLINE EXCLUSIVE PRICE
    1806.00 NOK
    1033.82 NOK / Pack of 6
    Save 772.18 NOK
    42% Off
Estimated Shipment: 07-01-2027
Add to basket
Add to basket
For shipping test tubes containing infectious substances or diagnostic specimens
  • Bubble-wrap compartment
  • Liquid-tight, plastic secondary container withstands pressure up to 95kPa
  • U.N. markings and a label for both infectious substances and diagnostic specimens
Compliance
Complies with ICAO/IATA Packing Instruction 650
Specifications
Certifications/Compliance Packing instruction for UN 3373/Category B label is PI 650
Applications For shipping test tubes containing infectious substances or diagnostic specimens
Size 4 x 4 x 8 in.
Type Infecon 2000 Shipper