Learn More
Fisherbrand™ Gel-Loading Aerosol-Barrier Tips
GREENER_CHOICE
Product Code. 11967734 Shop All Fisherbrand Products
Change view
Click to view available options
Volume (Metric):
10 to 100 μL
20 μL
Packaging:
Racked
Unit Size:
Pack of 1020
Product Code. Volume (Metric) Packaging unitSize
11967734 10 to 100 μL Racked Pack of 1020
12992571 20 μL Racked Pack of 1020
2 options
Product Code. 11967734
Only null left
Add to basket
Add to basket
Help minimize liquid retention
  • No RNase or DNase contamination
  • Natural color
  • For use with, but not limited to: Biohit Proline™ Electronic, BrandTech™ Transferpette™, Eppendorf™ Reference 2000, Eppendorf Research 2100, Thermo Finnpipette™ Focus, Fisherbrand™ Finnpipette™, Fisherbrand Finnpipette II, Genemate™ ISC Bio Express, Hamilton SoftGrip™, Jencons Sealpette™ Electronic, Labnet Labpette™ Digital, Mettler Toledo VoluMate™, Nichiryo 5000, Oxford™ Benchmate™, Pipetman™, Rainin™, Vistalab Ovation
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 10 to 100 μL
Filtered Filtered
Sterility Sterile
Color Natural
Length (English) 2.7 in.
Length (Metric) 69 mm
For Use With (Application) Help minimize liquid retention
Format Rack
Tip Style Gel Loading
Packaging Racked