Description
Extremely low plunger forces work longer without fatigue and reduce the risk of RSI.
- Fully autoclavable — entire pipette can be steam autoclaved
- Soft-touch tip ejection — a gearing mechanism reducing tip ejection force up to 50%
- Ergonomic comfortable lightweight handle — comfortable grip and effortless tip attachment
- Optimal fit with SureOne™ tips — best pipetting performance
- Super blow-out piston — creates 150% bigger air push than conventional piston for delivery of the micro size drops
Garantie
Three years.
Spécification
Spécification
| Accuracy | 0.6 to 1% |
| Compatible Tips | SureOne™ tips |
| Autoclavable | Autoclavable |
| For Use With | SureOne™ Tip |
| Product Line | Elite |
| Type | Micro Pipette |
| Volume (Metric) | 100 to 1000 μL |
| Finger Style | Ergonomic |
| Increments | 1μL |
| Secure Tip Attachment | Soft touch tip ejection |
| Afficher plus |
As part of our commitment to continuous product improvement, your delivery may contain products with some minor color variation, but their form, fit and function will not be affected. Any questions contact your sales representative.
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.