Learn More
Fisherbrand™ Disposable Borosilicate Glass Tubes with Threaded End
Shop All Fisherbrand Products
Click to view available options
Capacity (Metric):
8 mL
12 mL
16 mL
20 mL
25 mL
30 mL
36 mL
Length (Metric):
100 mm
125 mm
150 mm
Label Type:
Marking Spot
None
Thread Size:
13-415
15-415
18-415
Unit Size:
Pack of 1000
Pack of 500
Specifications
Specifications
| Certifications/Compliance | ASTM E438, Type I, Class B |
| Capacity (Metric) | 8 mL |
| Length (Metric) | 100 mm |
| Material | Borosilicate Glass |
| Disposable | Single-use |
| For Use With | For tissue culture work and general bacteriological use |
| Label Type | Marking Spot |
| Packaging | Tray |
| Thread Size | 13-415 |
| Sterility | Non-sterile |
| Show More |