Learn More
Fisherbrand™ Disposable Borosilicate Glass Tubes with Polypropylene Screw Cap
Product Code. 11577413
Change view
Click to view available options
Length (Metric):
100 mm
125 mm
150 mm
Diameter (Metric):
13 mm
16 mm
20 mm
Quantity:
1000 Pack
250/Tray
500/Tray
Unit Size:
Pack of 1000
Pack of 500
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Diameter (Metric) Quantity Unit Size
11577413 150 mm 20 mm 250/Tray Pack of 500
11527413 100 mm 16 mm 1000 Pack Pack of 1000
10421541 125 mm 16 mm 250/Tray Pack of 1000
11557413 150 mm 16 mm 250/Tray Pack of 1000
11567413 125 mm 20 mm 250/Tray Pack of 500
11517413 100 mm 13 mm 500/Tray Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 11577413
Estimated Shipment: 12-01-2027
Add to basket
Add to basket
Provides chemical stability and biological inertness of hard, borosilicate glass at price of disposable tubes
  • Natural linerless polypropylene screw cap
  • With marking spot
Ideal for tissue culture, bacteriology, clinical chemistry, blood typing, and crossmatching procedures
Order Info
Packed in trays of 250 or 500
Specifications
Capacity (Metric) 30 mL
Length (Metric) 150 mm
Material Borosilicate Glass
Disposable Single-use
For Use With Provides chemical stability and biological inertness of hard, borosilicate glass at price of disposable tubes
Label Type Marking Spot
Packaging Tray
Quantity 250/Tray
Sterility Non-sterile
Diameter (Metric) 20 mm
Closure Type Screw Cap
Product Type Disposable Culture Tube