Learn More
Fisherbrand™ Disposable Aluminum Dishes with Fluted Sides
Product Code. 11774059 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Capacity (Metric):
20 mL
60 mL
75 mL
200 mL
500 mL
Diameter (Metric) Inner Top:
43 mm
62 mm
64 mm
109 mm
180 mm
Diameter (Metric) Inner Base:
40 mm
46 mm
59 mm
77 mm
137 mm
Unit Size:
Pack of 1000
Pack of 1200
Pack of 50
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Diameter (Metric) Inner Top Diameter (Metric) Inner Base Unit Size
11774059 200 mL 109 mm 77 mm Pack of 50
11729418 20 mL 43 mm 40 mm Pack of 1000
11739418 60 mL 64 mm 59 mm Pack of 1200
11749418 75 mL 62 mm 46 mm Pack of 1000
11784059 500 mL 180 mm 137 mm Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Artikelnummer. 11774059
Top Sellers. Mega Savings.
Use promo code 27997
Alle Werbeaktionen anzeigen
In Stock
Zum Warenkorb hinzufügen
Edge
Zum Warenkorb hinzufügen
Useful as evaporating and general utility dishes
Fisherbrand™ Disposable Aluminum Dishes with Fluted Sides are inexpensive enough for single use; useful as evaporating and general utility dishes
Larger sizes are ideal for food processing applications.
  • Made of lightweight aluminum
  • Flat bottoms
  • Fluted or slightly fluted sides
  • Flanged rims
Spezifikation
Bottom Shape Flat
Capacity (Metric) 200 mL
Disposable Single-use
Material Aluminum
Shape Round
Type Weighing Dish
For Use With (Application) Useful as evaporating and general utility dishes
Diameter (Metric) Inner Base 77 mm
Diameter (Metric) Inner Top 109 mm
Diameter (Metric) 109 (Inner Top) mm, 77 mm (Inner Base)