Learn More
Fisherbrand™ Bitran™ Specimen Storage Bags
Product Code. p-32660 Shop All Fisherbrand Products
Change view
Click to view available options
Dimensions (L x W):
2 x 4 in. (5.1 x 10.2cm)
Material:
Saranex Film
Length (Metric):
5.1 cm
Width (Metric):
10.2 cm
Unit Size:
Pack of 250
Product Code. Dimensions (L x W) Material Length (Metric) Width (Metric) unitSize
16488115 2 x 4 in. (5.1 x 10.2cm) Saranex Film 5.1 cm 10.2 cm Pack of 250
1 options
Product Code. 16488115
Only null left
Add to basket
Add to basket
Choice of Saranex™ odor-control film for long-term storage or polyethylene for short-term storage
  • Use for collecting, transporting and storing tissue specimens
  • Minimize storage space and help to eliminate leakage, breakage and contamination
  • Resealable double-lock zipper closure
  • Designed for storage of tissues and fixatives and for use as mini-anaerobic environments
Specifications
Closure Type Zip Seal
Material Saranex Film
Quantity 250
Dimensions (L x W) 2 x 4 in. (5.1 x 10.2cm)
Length (English) 2 in.
Length (Metric) 5.1 cm
Width (English) 4 in.
Width (Metric) 10.2 cm