Learn More
Fisherbrand™ Bulk Beads
Product Code. 15505809 Shop All Fisherbrand Products
Change view
Click to view available options
Particle Size:
1.4 mm
2.38 mm
2.8 mm
Unit Size:
1 pack
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Particle Size unitSize
15505809 2.38 mm 1 pack
15515809 1.4 mm 1 pack
15525809 2.8 mm 1 pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15505809
Estimated Shipment: 04-09-2026
Add to basket
Add to basket
For use with Bead Mill 4 and 24 Homogenizers
  • 500g of beads to fill up to 500 x 2mL sample tubes
  • Sample tubes not included
TRUSTED_SUSTAINABILITY
Specifications
Product Type Metal Bulk Beads
Particle Size 2.38 mm
Material Metal
For Use With (Equipment) Bead Mill 4 and 24 Homogenizers
Packaging Quantity 500 g