Specifications
| Capacity (English) | 4 oz. |
| Capacity (Metric) | 120 mL |
| Closure Type | Round Wire |
| Color | Clear |
| Material | Polyethylene |
| Sterility | Sterile |
| Thickness (English) | 3.50 mil |
| Dimensions (L x W) | 7 x 3 in. |
| Length (English) | 7 in. |
| Length (Metric) | 17.8 cm |
| Show More |