Learn More
Fisherbrand™ Aluminum Weighing Dishes
Product Code. 10297593 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Capacity (Metric):
20 mL
42 mL
65 mL
70 mL
150 mL
Depth (Metric):
13 mm
16 mm
17 mm
17.3 mm
20 mm
Diameter (Metric) Inner Base:
44 mm
51 mm
68.2 mm
69 mm
96 mm
Shape:
Flanged Sides
Fluted Sides
Unit Size:
Pack of 100
Pack of 1000
Pack of 144
Pack of 8064
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Depth (Metric) Diameter (Metric) Inner Base Shape Unit Size
10297593 42 mL 17 mm 51 mm Fluted Sides Pack of 144
11799408 65 mL 16 mm 69 mm Fluted Sides Pack of 1000
11704069 42 mL 17 mm 51 mm Fluted Sides Pack of 8064
11789408 20 mL 13 mm 44 mm Fluted Sides Pack of 1000
11588372 20 mL 13 mm 44 mm Fluted Sides Pack of 100
11709418 70 mL 17.3 mm 68.2 mm Fluted Sides Pack of 1000
11518382 150 mL 20 mm 96 mm Flanged Sides Pack of 100
11719418 150 mL 20 mm 96 mm Flanged Sides Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 10297593
Estimated Shipment: 18-12-2026
Add to basket
Add to basket
Aluminum dishes for weighing and dispensing
Choose Fisherbrand™ Aluminum Weight Dishes for weighing and dispensing.
With Fluted sides
  • Also can serve as evaporating dishes or dust covers Lightweight, yet sturdy Convenient finger-grip handle can be etched prior to weighing for identification
With Smooth sides
  • Lightweight dishes with flanged rims
  • Slightly sloping sides facilitate stacking
  • Especially useful for materials that cannot easily be cleaned from dish surface
Specifications
Bottom Shape Flat
Capacity (Metric) 42 mL
Material Aluminum
Shape Fluted Sides
Type Weighing Dish
For Use With (Application) For weighing and dispensing
Depth (Metric) 17 mm
Diameter (Metric) Inner Base 51 mm
Diameter (Metric) Inner Top 62 mm
Diameter (Metric) 51 mm (Inner Base), 62 mm (Inner Top)