Learn More
Fisherbrand™ Aluminum Foil, Standard-Gauge Roll
Product Code. 11759408 Shop All Fisherbrand Products
Click to view available options
Length (English):
25 ft.
197 ft.
492 ft.
Thickness:
18 μm
24 μm
Width (Metric):
30cm
30.5cm
46cm
Length (Metric):
150 m
60 m
7.6 m
Unit Size:
Each
Product Code. 11759408
Only null left
Add to basket
Edge
Add to basket
For sealing, weighing, and transferring samples
Ideal for weighing, sealing, and tranferring your samples.
  • Packed in individual dispenser box with cutter bar
  • Heavy-gauge rolls offer greater folding strength, good for covering sharp edges
Specifications
Applications Sealing, Weighing, Transferring Samples
Length (English) 25 ft.
Product Type Foil
Width (English) 11.9 in.
Width (Metric) 30cm
Length (Metric) 7.6 m
Material Aluminum
Thickness 18 μm