Learn More
Fisherbrand™ 96-Well Polystyrene Plates
Product Code. 13525440 Shop All Fisherbrand Products
Change view
Click to view available options
Color:
Black
Clear
White
Well Shape:
Flat Bottom (F-well)
Round Bottom (U-well)
Unit Size:
Pack of 180
Pack of 80
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Well Shape unitSize
13525440 White Flat Bottom (F-well) Pack of 180
11997944 Clear Flat Bottom (F-well) Pack of 80
11987944 Clear Round Bottom (U-well) Pack of 80
13535440 Black Flat Bottom (F-well) Pack of 180
13575480 Clear Round Bottom (U-well) Pack of 180
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 13525440
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Ideal for antibiotic screens, cell-based assays and screening compounds.
Ideal for antibiotic screens, cell-based assays and screening compounds
Designed for compatibility with automated equipment
  • Polystyrene
  • Plates stack easily without lids
  • Available in flat or round well shapes
  • Nonsterile
  • Untreated, provides uniform surface properties
  • Use 12-565-511 lid (sold separately)
TRUSTED_SUSTAINABILITY
Specifications
Format Standard
Array 8 x 12
Color White
Material Polystyrene
Volume (Metric) Well 400 μL
No. of Wells 96
Volume (Metric) Working 50-250 μL/well
Sterility Non-sterile
Surface Treatment Non-treated
Well Shape Flat Bottom (F-well)
For Use With (Application) Ideal for antibiotic screens, cell based assays and screening compounds
Quantity 180/Cs.