Learn More
Fisherbrand™ 384-Well ShallowWell Polypropylene Microplates
Product Code. 13585450 Shop All Fisherbrand Products
Change view
Click to view available options
Color:
Black
White
Sterility:
Non-sterile
Sterile
Volume (Metric) Well:
58 μL
120 μL
For Use With (Application):
Fluorescent Assays
Luminescent Assays
Storage or Colorimetric Assays
Unit Size:
Pack of 100
Product Code. Color Sterility Volume (Metric) Well For Use With (Application) unitSize
13585450 White Non-sterile 120 μL Luminescent Assays Pack of 100
13565450 White Non-sterile 120 μL Storage or Colorimetric Assays Pack of 100
13575450 White Sterile 58 μL Storage or Colorimetric Assays Pack of 100
13595450 Black Non-sterile 120 μL Fluorescent Assays Pack of 100
4 options
Product Code. 13585450
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Small volume applications
Feature a shared-wall design which yields increased well volume
  • Automation compatible, ANSI standard
  • Low-binding and chemically resistant surface
  • Natural plates for storage or colorimetric assays
  • White plates for luminescent assays, including SPA
  • Black plates for fluorescent assays
  • Round bottom wells for optimal sample recovery
  • Rounded square wells minimize wicking (capillary action)
  • Working volume range: 2 to 35μL/well
Order Info
Lids not included
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ANSI
Format Standard
Array 16 x 24
Color White
Material Polypropylene
Volume (Metric) Well 120 μL
No. of Wells 384
Sterility Non-sterile
Well Shape Round, Conical Bottom (V-well)
For Use With (Application) Luminescent Assays