Learn More
Fisherbrand™ Vortex Mixer Accessories: Foam Insert Kit
For use with Fisherbrand vortex mixers.
Brand: Fisherbrand™
Additional Details : Weight : 0.10000kg
Description
Also compatible with other manufacturers' models; contact your Fisher Scientific Customer Service Representative for details.
Foam Insert Variety Pack
- Includes foam inserts for the following: 9 to 13mm tube holder, 14 to 19mm tube holder, 20 to 29mm
Specifications
Blank Foam insert | |
Foam | |
Vortex mixer |