Learn More
Fisherbrand™ Traceable™ Relative Humidity/Temperature Meters
Product Code. 11765843 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Type:
Dig. Humidity/Temp Mete w/o Memory
Dig. Humidity/Temp Meter w/ Readout
Digital Humidity Meter
Height (Metric):
10 cm
18 cm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Height (Metric) Unit Size
11765843 Dig. Humidity/Temp Meter w/ Readout 10 cm Each
11764186 Digital Humidity Meter 10 cm Each
11774186 Dig. Humidity/Temp Mete w/o Memory 18 cm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11765843
Spotlight Offers
Use promo code 28037
View all promotions
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket
Monitor air conditions in environmental chambers, chemical storage areas, cleanrooms, incubators and stockrooms
Fisher Scientific™ Traceable™ Relative Humidity/Temperature Meters allow you to monitor air conditions in environmental chambers, chemical storage areas, cleanrooms, incubators and stockrooms.
  • Humidity range: 25 to 95% RH, Accuracy: ±3%RH (25 to 75)%RH, ±5%RH otherwise
  • Continuously and simultaneously display relative humidity (11-661-11) or humidity/temperature (11-661-12) in extra-large, 0.62 in.-high (1.5 cm) digits
  • Made from high-impact, chemical-resistant ABS plastic
With Temperature Readout
  • Temperature readout in °C or °F; ranges from 0° to 50°C (32° to 122°F) 
  • Measures 0.75L x 3.25W x 7 in.H (2 x 8 x 18 cm)
  • Net weight: 5 oz. (142g)
  • Wall mount included
With Temperature Readout and Dual Min./Max. Memory
  • Temperature readout in °C and °F; ranges from 0° to 50°C (32° to 122°F) 
  • Minimum and maximum memories permit air monitoring overnight, during weekends, or for virtually any other time period
  • Measures 0.62L x 2.62W x 3.87 in.H (2 x 7 x 10 cm)
  • Net weight: 2 oz. (57g)
  • Wall mount and bench stand included
Certifications
A certificate is provided to indicate instrument traceability to standards provided by the National Institute of Standards and Technology (NIST™). Calibrated by an ISO™ 17025 calibration laboratory accredited by A2LA™.
Compliance
Cat. No. 11-661-13 meets federal requirements for CLIA Specification 493.1207 for monitoring temperature and humidity in clinical laboratories.
Specifications
Type Dig. Humidity/Temp Meter w/ Readout
Humidity Range 25 to 95%
Resolution 0.01
Accuracy ±3%RH (25 to 75)%RH, ±5%RH otherwise
Temperature Range (Metric) 0°C to 50°C
Temperature Accuracy ±1°C (0 to 40°C) otherwise ±2°C
Display Type LCD
Material ABS Plastic
Memory Stores and recalls minimum/ maximum temperature and humidity readings, display up-dated every second
Sensors Solid State
Temperature Range (English) 32°F to 122°F
Temperature Resolution 1°F
Dimensions (L x W x H) 0.62 x 2.62 x 3.87 in. (1.57 x 7 x 10 cm)
Length (English) 0.62 in.
Length (Metric) 1.57 cm
Width (English) 2.62 in.
Width (Metric) 7 cm
Height (English) 3.87 in.
Height (Metric) 10 cm
Weight (English) 2.5 oz.
For Use With (Application) Monitor air conditions in environmental chambers, chemical storage areas, cleanrooms, incubators and stockrooms
Includes Bench stand, switch to +read °F/°C, fast-response solid-state sensors and two AAA batteries
Certifications/Compliance ISO 17025, A2LA, NIST