Learn More
Fisherbrand™ Traceable™ Digital Thermometers with Short Sensors
Product Code. 11735843 Shop All Fisherbrand Products
Click to view available options
Temperature Range (Metric):
-5°C to +50°C
-50°C to +70°C
Temperature Range (English):
-58°F to +158°F
23°F to 122°F
N/A
Scale:
Celsius
Celsius, Fahrenheit
Length (Metric):
57 mm
70 mm
102 mm
108 mm
Unit Size:
Each
Product Code. 11735843
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Fisherbrand™ Traceable™ Digital Thermometers are ideal for monitoring liquids, air/gas or semisolids.
Choose from models with a variety of features including MAX/MIN function to record the maximum and minimum temperature in a certain time period; visual and audible alarms and multiple displays of recorded temperatures.
  • Select the correct temperature to use in water baths, refrigerators, incubators and tanks
Certifications
Individually serial-numbered Traceable Certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA™ indicates traceability to standards provided by NIST™.
Specifications
Certifications/Compliance ISO 17025, A2LA, NIST
Diameter (English) Probe 0.18 in.
Dimensions (L x W x H) 2.25 x 3.25 x 2.25 in. (5.7 x 8.2 x 5.71 cm)
For Use With (Application) Monitoring liquids, air/gas or semisolids in water baths, refrigerators, freezers, incubators and tanks
Height (English) 2.25 in.
Height (Metric) 5.71 cm
Includes Sensor, AAA alkaline battery, flip open stand, wall mount, magnet and Velcro™ mounting tape
Length (English) Cable 10 ft.
Length (English) Probe 0.8 in.
Length (Metric) 57 mm
Length (Metric) Cable 305 cm
Length (Metric) Probe 2.03 cm
Material Plastic
Resolution 0.1°C
Weight (English) 2.5 oz.
Weight (Metric) 70.8 g
Width (English) 3.25 in.
Length (English) 2.25 in.
Max. Temperature (English) 158°F
Max. Temperature (Metric) 70°C
Min. Temperature (English) -58°F
Min. Temperature (Metric) -50°C
Temperature Range (English) -58°F to +158°F
Temperature Range (Metric) -50°C to +70°C
Calibrated Yes
Accuracy ±1°C
Display Digital LCD
Width (Metric) 8.2 cm
Scale Celsius, Fahrenheit