Learn More
Fisherbrand™ Cup Horn for Model 120 Sonic Dismembrator
Product Code. 12991181 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Diameter (English):
1.5 in. Cup horn
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (English) Unit Size
12991181 1.5 in. Cup horn Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12991181
  • ONLINE EXCLUSIVE PRICE
    9640.00 NOK
    9115.75 NOK / Pack of 1
    Save 524.25 NOK
    5% Off
Estimated Shipment: 21-12-2026
Add to basket
Add to basket
1.5 in. Diameter Cup Horn for Model 120 Sonic Dismembrator. Process samples indirectly to prevent cross contamination.
The Fisher Scientific™ Cup Horn for the Model 120 Sonic Dismembrator is used for indirect processing of individual sealed tubes or vials.
  • Indirect sonication accessory to process one sample at a time
  • Prevent cross contamination
  • Eliminate splashing and aerosols
  • Includes: 1.5 in. (3.8cm) Cup horn and 1.5ml tube holders.
Specifications
Product Type Cup Horn
Diameter (English) 1.5 in. Cup horn
For Use With (Application) Ideal for indirect processing of individual sealed tubes or vials
For Use With (Equipment) Model 120 sonic dismembrator
Holds 1 sample Tube
Includes 1.5 mL tube holder