Learn More
ROBU VitraPOR™ Filter Disc, Surfaces Plane, Ground Cylindrical, 280 mm
The faces of the VitraPOR™ biplane filter discs from series 16 are precisely ground down to tight tolerances and plane surfaces.
Brand:  ROBU VitraPOR™ 62802
Additional Details : Weight : 1.97600kg
 View more versions of this product
Product Code. 16883276
  • 12415.00 NOK / Each
Estimated Shipment: 14-06-2024
Description
  • Highly resistant to most chemicals and temperatures up to 500°C
  • The biplane version may be required in specific installation circumstances
Specifications
Specifications
Borosilicate Glass (3.3)
2
280 mm
Non-sterile
18.0 mm ± 0.5 mm