missing translation for 'onlineSavingsMsg'
Learn More

ROBU VitraPOR™ Filter Disc, Surfaces Plane, Ground Cylindrical, 140 mm

The faces of the VitraPOR™ biplane filter discs from series 16 are precisely ground down to tight tolerances and plane surfaces.

Brand:  ROBU VitraPOR™ 61404

Additional Details : Weight : 0.20900kg

 View more versions of this product

Product Code. 16893206

  • 2430.00 NOK / Each
Estimated Shipment: 03-06-2024
to see stock.
Add to basket

Description

Description

  • Highly resistant to most chemicals and temperatures up to 500°C
  • The biplane version may be required in specific installation circumstances
Specifications

Specifications

Borosilicate Glass (3.3)
1 Ea.
8.5 ± 0.2 mm
Non-sterile
4
140 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers