Learn More
Sartorius Stedim Biotech™ Filter Boards, Grade C 350L
Very pure filter paper made of cotton linters only and suitable for analytical sample preparation.
Brand: Sartorius Stedim Biotech™ FT-2-340-375500
Additional Details : Weight : 1.00000kg
Description
- Very pure paper made of cotton linters only
- Suitable for analytical sample preparation
- Non-sterile, bulk packed
Specifications
360 g/m2 | |
100% Cotton Linters (>98% alpha-cellulose) | |
100/Pk. | |
375 mm × 500 mm |
Sheet | |
C 350L | |
0.75 mm |